Description
- Blue Live Cell Caspase Probe is excited by the blue laser (with an excitation maximum of 492nm), and has a fluorescence emission maximum of 514nm
Specifications
Specifications
| Quantity | 50 Tests |
| Reactivity | Human, Mouse |
| Product Type | Blue Live Cell Caspase Probe |
| Content And Storage | -20°C |
| For Use With (Application) | Flow Cytometry, Intracellular Staining(Flow Cytometry) |