Learn More
BD Pharmingen™ IHC Zinc Fixative
Product Code. 10400275
Change view
Click to view available options
Quantity:
500mL
1000mL
Dilution:
1 X
10 X
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dilution unitSize
10400275 1000mL 1 X Each
15886838 500mL 10 X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10400275 Supplier BD Supplier No. 550523
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
An alternative to formalin fixation of tissue for paraffin embedded sections
Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.
  • Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
  • Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases
There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Content And Storage Room temperature
Dilution 1 X
Product Type Fixative
Quantity 1000mL
For Use With (Application) Immunohistochemical staining
Form Liquid