Learn More
BD DRAQ7™
Product Code. 15813959
Change view
Click to view available options
Quantity:
1mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15813959 1mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15813959 Supplier BD Supplier No. 564904
 Restricted Item
Estimated Shipment: 30-10-2026
Used to stain nucleic acids in fixed cells. BD Pharmingen™ DRAQ7™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or neucleated cells.
  • Far-red fluorescent DNA dye
  • Cell impermeable
  • Used to stain nucleic acids in fixed cells
  • Has an excitation wavelength maxima of 599/644nm
TRUSTED_SUSTAINABILITY
Specifications
Color Far-red
Concentration 0.3mM
Content And Storage Store undiluted at 4°C
Excitation Wavelength Range 599/644nm
Dye Type DNA dye
For Use With (Application)
  • Staining of Live Cells for Viability Analysis by Flow Cytometry
  • Staining of Fixed Cells for DNA Content Analysis by Flow Cytometry
  • Immunofluorescent Staining of Fixed Cells for Nuclear Visualization
Reactivity
  • Tested in Development: Human, Mouse
  • Reported Reactivity: Rat
Quantity 1mL
compliance-icons
Product Identifier
  • DRAQ7 1.0ml
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-BIS{[2-(DIMETHYLAMINO) ETHYL]AMINO}-4,8-DIHYDROXYANTHRACENE-9,10-DIONE