Learn More
BD DRAQ5™
Product Code. 15803959
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15803959 50 μL Each
15893949 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15803959 Supplier BD Supplier No. 564903
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
Used to stain nucleic acids in live or fixed cells. BD Pharmingen™ DRAQ5™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or enucleated cells.
  • Far-red fluorescent DNA dye
  • Cell permeable
  • Used to stain nucleic acids in live or fixed cells
  • Has an excitation wavelength maxima of 598/646nm
TRUSTED_SUSTAINABILITY
Specifications
Color Far-red
Concentration 5 mM
Content And Storage Store undiluted at 4°C
For Use With (Application) Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry)
Dye Type DNA dye
Excitation Wavelength Range 598/646 nm
Quantity 50 μL
Reactivity Tested in Development: Human, Mouse; Reported Reactivity: Rat