Learn More
BD Yellow-Green Live Cell Caspase Probe
Product Code. 15864369 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864369 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15864369 Supplier BD Supplier No. 565520
 Restricted Item
Estimated Shipment: 18-11-2026
Cells with active caspases can be detected. BD Pharmingen™ Yellow-Green Live Cell Caspase Probe contains a fluorochrome-labeled caspase inhibitor. Fluorochrome-labeled caspase inhibitors have three functional domains: fluorochrome , 3-amino acid sequence (VAD) that binds to the active site of the activated caspase, and fluoromethyl ketone (FMK) moiety that allows the probe to irreversibly bind the active caspase.
  • Yellow-Green Live Cell Caspase Probe is excited by the yellow-green laser (with an excitation maximum of 556nm), and has a fluorescence emission maximum of 575nm
  • The probe is also well-excited by blue laser
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 Tests
Reactivity Human, Mouse
Product Type Yellow-Green Live Cell Caspase Probe
Content And Storage -20°C
For Use With (Application) Flow Cytometry, Intracellular Staining(Flow Cytometry)