Learn More
BD PBX Calcium Assay Kit
Product Code. 15816289
Change view
Click to view available options
Quantity:
10 Preps
100 Preps
Unit Size:
1 set
Product Code. Quantity unitSize
15816289 100 Preps 1 set
15896279 10 Preps 1 set
2 missing translation for 'options'
Product Code. 15816289
Brand: BD 640177
Includes
Calcium Indicator, 10X PBX Signal Enhancer
For homogeneous measurement of intracellular calcium changes
  • Involves one dye addition step
  • Does not require washing
  • Allows easy implementation in highthroughput environment
  • Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
  • Can improve retention of the dye in cells
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Calcium indicator at -20°C and PBX Signal Enhancer 10X at room temperature
For Use With (Application) For homogeneous measurement of intracellular calcium changes
Includes 10 vials calcium indicator,100mL PBX signal enhancer (10X, lyophilized)
Quantity 100 Preps
Type Calcium Assay Kit