missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD OptEIA™ Mouse IL-6 ELISA Kit
Description
Includes
Antibody coated plates, detection antibody, standards, streptavidin-HRP, diluents, wash buffer, TMB substrate, stop solution, plate sealers.
- Solid phase sandwich ELISA
- Uses a monoclonal antibody specific for IL-6 coated on a 96-well plate
- Size: 2 Plate(s)
Specifications
Specifications
| Antigen | IL-6 |
| Cross Reactivity | Mouse |
| Includes | Wash concentrate (20X), stop solution, Enzyme concentrate Streptavidin HRP, ELISA diluent III, standard/Sample diluent, TMB one-step Substrate reagent, Mouse IL-6 Antibody coated plate, detection Antibody Biotin Anti-Mouse IL-6, Mouse IL-6 standard |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Quantitative determination of Mouse IL-6, ELISA |
| Product Type | Mouse IL-6 ELISA kit |
For research use only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?