Learn More
BD Luer-Lok™ Disposable Syringes without Needles
Click to view available options
Volume (Metric):
5 mL
20 mL
30 mL
Graduations:
0.2 mL
1 mL
Connection Type:
Luer Lock
Slip Tip
Unit Size:
1 Each
Pack of 100
Pack of 40
Description
PrecisionGlide™ and Yale™ disposable and reusable needles are available
- Supplied with tip shield
- All syringes are for general purposes
- 14-827-52 control syringe
- Slip tip, Luer-Lok™ tip/cap available separately
Syringes with two capacities listed are dual scale.
Specifications
Specifications
| Graduations | 1 mL |
| Needle Point Number | No Needle Included |
| Volume (Metric) | 30 mL |
| Connection Type | Luer Lock |