Learn More
BD Human IL-8 ELISA Kit II
Product Code. 15866468
Click to view available options
Quantity:
2 Plates
Unit Size:
1 set
Product Code. 15866468
Brand: BD 550999
Includes
Antibody coated wells, Detection antibody, Standards, Enzyme Concentrate (250x), Standard/Sample Diluent, ELISA Diluent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers
Developed for superior accuracy with serum and plasma specimens
Helps in the quantitative determination of human IL-8 in serum, plasma, and cell culture supernatant.
  • Related to low molecular weight proinflammatory factors known as chemokines
Applications:
ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen IL-8
Cross Reactivity Human
Includes Wash concentrate (20X), stop solution, Enzyme concentrate Streptavidin HRP, Human IL-8 Antibody coated plate, detection Antibody Biotin Anti-Human IL-8, recombinant Human IL-8 standard, standard/Sample Diluent, TMB one-step Substrate reagent, ELISA Diluent
Quantity 2 Plates
Sample Type Serum, Plasma
Content And Storage 2°C to 8°C
For Use With (Application) Quantitative determination of Human IL-8, ELISA
Product Type Human IL-8 ELISA kit II
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BD OptEIA Human IL-8 ELISA Kit II
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : phosphoric acid, CMIT/MIT, 5-chloro-2-methyl-4-isothiazolin-3-one