missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IL-12 (p70) ELISA Kit
Click to view available options
Quantity:
2 Plates
Unit Size:
1 set
Description
Includes
2 plates of 96 breakable wells coated with anti-human IL-12 antibody; Detection antibody: Biotinylated anti-human IL-12; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-12, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
- Contains biologically active p70 which stimulates the growth of activated CD4+ and CD8+ T cells and NK cells
Specifications
Specifications
| Antigen | IL-12 (p70) |
| Cross Reactivity | Human |
| Includes | 2 Plates of 96 breakable well coated with Anti-Human IL-12 Antibody, Biotinylated Anti-Human IL-12, HRP Conjugate (250X), 4 vials recombinant Human IL-12 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | In-vitro quantitative determination of Human IL-12 (p70), ELISA |
| Product Type | Human IL-12 (p70) ELISA kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction