Learn More
BD Human IL-10 ELISA Kit II
Click to view available options
Quantity:
2 Plates
Unit Size:
1 set
Description
Includes
2 plates of 96 breakable wells coated with anti-human IL-10 antibody; Detection antibody: Biotinylated anti-human IL-10; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-10, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
The OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).
- Reactivity: Human
- Utilizes monoclonal antibody specific for IFN-γ coated on a 96-well plate
Applications:
ELISA
Specifications
Specifications
| Antigen | IL-10 |
| Cross Reactivity | Human |
| Includes | 2 Plates of 96 breakable well coated with Anti-Human IL-10 Antibody, Biotinylated Anti-Human IL-10, HRP Conjugate (250X), 4 vials recombinant Human IL-10 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Quantitative determination of Human interleukins, ELISA |
| Product Type | Human IL-10 ELISA kit II |