missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Rat Anti-Mouse CD25

Product Code. 15884759
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15884759

Brand: BD 566120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CD25 is transiently expressed at a low level during normal B-cell development in the bone marrow on the CD45R/B220low TdT- sIg- Pre-B/Pre-B-II and CD45R/B220low TdT- sIgM+ sIgD- immature B stages. BD Horizon™ BV480 Rat Anti-Mouse CD25 is expressed at a higher level during a very early stage of T-cell development in fetal and adult thymus.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Mouse CD25
Quantity 0.1 mg
For Use With (Application) Flow Cytometry
Concentration 0.2 mg/mL
Cross Reactivity QC Testing: Mouse
Form BV480
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.