Learn More
BD BV480 Mouse Anti-Human CD279 (PD-1)
Product Code. p-6909023
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
Product Code. 15804819
Brand: BD 566175
CD279 is an immunoregulatory receptor expressed on activated T cells, B cells, and myeloid cells. BD Horizon™ BV480 Mouse Anti-Human CD279 (PD-1) contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms.
With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications
Product Type BV480 Mouse Anti-Human CD279 (PD-1)
Quantity 25 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Gene ID (Entrez) 5133
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide