Learn More
BD BV480 Mouse Anti-Human CD14
Product Code. 15864779
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864779 100 Tests Each
15834829 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864779 Supplier BD Supplier No. 566141
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. BD Horizon™ BV480 Mouse Anti-Human CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells.
  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications
Product Type BV480 Mouse Anti-Human CD14
Quantity 100 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide