Learn More
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Product Code. p-3201178
Change view
Click to view available options
Product Type:
Salmonella O Antiserum Factor 10, Group E1
Salmonella O Antiserum Factor 14
Salmonella O Antiserum Factor 15, Groups E2 and E3
Salmonella O Antiserum Factor 19, Group E4
Salmonella O Antiserum Factor 2
Salmonella O Antiserum Factor 20
Salmonella O Antiserum Factor 22, Group G1
Salmonella O Antiserum Factor 23, Group G2
Salmonella O Antiserum Factor 27
Salmonella O Antiserum Factor 34, Group E3
Salmonella O Antiserum Factor 4, Group B
Salmonella O Antiserum Factor 5, Group B
Salmonella O Antiserum Factor 8, Groups C2 and C3
Salmonella O Antiserum Factor 9, Group D
Salmonella O Antiserum Factors 4, 5, Group B
Salmonella O Antiserum Group D1 Factors 1, 9, 12
Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Salmonella O Antiserum Group E1 Factors 3, 10
Salmonella O Antiserum Group F Factor 11
Salmonella O Antiserum Group G Factors 13, 22
Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
Salmonella O Antiserum Group J Factor 17
Salmonella O Antiserum Group L Factor 21
Salmonella O Antiserum Group M Factor 28
Salmonella O Antiserum Group N Factor 30
Salmonella O Antiserum Group O Factor 35
Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
Salmonella O Antiserum Poly F, Groups 51-55
Salmonella O Antiserum Poly G, Groups 56-61
Förpackningsstorlek:
3mL
Each
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Product Type unitSize
11768153 Salmonella O Antiserum Group M Factor 28 3mL
11788153 Salmonella O Antiserum Group N Factor 30 3mL
11758153 Salmonella O Antiserum Group J Factor 17 3mL
17312361 Salmonella O Antiserum Group G Factors 13, 22 Each
17322361 Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25 Each
11742804 Salmonella O Antiserum Group O Factor 35 3mL
11792844 Salmonella O Antiserum Factor 14 3mL
11722804 Salmonella O Antiserum Group L Factor 21 3mL
11782904 Salmonella O Antiserum Group D1 Factors 1, 9, 12 3mL
11792734 Salmonella O Antiserum Factor 10, Group E1 3mL
11702744 Salmonella O Antiserum Factor 15, Groups E2 and E3 3mL
11712744 Salmonella O Antiserum Factor 19, Group E4 3mL
17302361 Salmonella O Antiserum Group F Factor 11 Each
11748153 Salmonella O Antiserum Factor 34, Group E3 3mL
11732814 Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H 3mL
11772804 Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L 3mL
11742814 Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O 3mL
11752814 Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U 3mL
11762814 Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z 3mL
11742844 Salmonella O Antiserum Poly F, Groups 51-55 3mL
11702854 Salmonella O Antiserum Factor 20 3mL
11772844 Salmonella O Antiserum Factor 4, Group B 3mL
11752844 Salmonella O Antiserum Poly G, Groups 56-61 3mL
11782844 Salmonella O Antiserum Factor 5, Group B 3mL
11752854 Salmonella O Antiserum Factor 23, Group G2 3mL
11742854 Salmonella O Antiserum Factor 22, Group G1 3mL
11772854 Salmonella O Antiserum Factor 27 3mL
11792874 Salmonella O Antiserum Factor 2 3mL
11702884 Salmonella O Antiserum Factors 4, 5, Group B 3mL
11722884 Salmonella O Antiserum Factor 8, Groups C2 and C3 3mL
11732884 Salmonella O Antiserum Factor 9, Group D 3mL
11792904 Salmonella O Antiserum Group E1 Factors 3, 10 3mL
17342361 Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Denna artikel kan inte returneras. Se returpolicy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Produktkod. 11768153 Leverantör BD Leverantörsnummer 211781
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative
TRUSTED_SUSTAINABILITY
Specifikationer
Certifications/Compliance For in vitro diagnostic use
Product Type Salmonella O Antiserum Group M Factor 28
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.