missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Violet Red Bile Agar

Product Code. p-4901939
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11758143

Brand: BD 211695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For enumerating coliform organisms in dairy products

  • Formulation: Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L

Order Info

Prices are for units of one each unless otherwise specified

Certifications

BAM, CCAM, COMPF, ISO, SMD

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF, ISO, SMD
Color Violet Red
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Violet Red Bile Agar
Product Type Agar
Formulation Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L
For Use With (Application) For enumerating coliform organisms in dairy products
Packaging Type Bottle
Quantity 500 g
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : oxgall
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.