missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ TC Yeastolate, ultra-filtered (UF)

Product Code. 16239951
Click to view available options
Quantity:
500 g
50 kg
Manufacturing Quality:
UF
Unit Size:
500g
50kg
This item is not returnable. View return policy

Product Code. 16239951

Brand: Gibco™ 670079

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Enhance cell culture performance with Gibco™ Difco™ TC Yeastolate Ultra-Filtered (UF) Low Bioburden while also mitigating risk in bioprocessing methods.

Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.

Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.

Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type TC Yeastolate
Format Drum
For Use With (Application) Preparing bacterial, insect, and mammalian cell culture media
Manufacturing Quality UF
Product Line Difco
Quantity 50 kg
Source Animal origin-free, yeast
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.