missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Phenol Red Mannitol Agar
Click to view available options
:
Phenol Red Mannitol Agar; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Phenol Red Mannitol Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone No. 3 10g/L, Beef extract 1g/L, D-Mannitol 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Phenol red 25mg/L |
| For Use With (Application) | Determination of ability of microorganisms to ferment mannitol |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction