missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Pantothenate Assay Medium

Product Code. 11729012
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11729012 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11729012 Supplier BD Supplier No. 260410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Microbiological assay of pantothenate

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Pantothenate Assay Medium
Product Type Pantothenate Assay Medium
Formulation Vitamin assay Casamino Acids 10g/L, Dextrose 40g/L, Sodium Acetate 20g/L, L-Cystine 0.4g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 200μg/L, Riboflavin 400
For Use With (Application) Microbiological assay of Pantothenate
Packaging Type Bottle
Quantity 100 g
compliance-icons
Product Identifier
  • panthothenate assay medium
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.