Learn More
BD Difco™ Dehydrated Culture Media: Oatmeal Agar
Click to view available options
:
Oatmeal Agar; 500g
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Oatmeal Agar |
| Product Type | Agar |
| Formulation | Oatmeal 60g/L, Agar 12.5g/L |
| For Use With (Application) | Cultivation of fungi, particularly for macrospore formation |
| Packaging Type | Bottle |
| Quantity | 500 g |