missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Listeria Enrichment Broth
Selective enrichment for L. monocytogenes from non-dairy and processed food products
117.00€
Specifications
Content And Storage | Refrigerate |
---|---|
Form | Dehydrated |
Media Type | Listeria Enrichment Broth |
Product Type | Microbiology Media |
Formulation | Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L |
Description
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Refrigerate | |
Listeria Enrichment Broth | |
Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L |
Dehydrated | |
Microbiology Media | |
Selectively enrich Listeria from food |