Learn More
BD Difco™ Dehydrated Culture Media: Listeria Enrichment Broth
Selective enrichment for L. monocytogenes from non-dairy and processed food products
Brand: BD 222220
Additional Details : Weight : 0.80000kg
Description
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
CE | |
Dehydrated | |
Microbiology Media | |
Selectively enrich Listeria from food | |
500 g |
Refrigerate | |
Listeria Enrichment Broth | |
Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L | |
Bottle |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : cycloheximide