missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Fraser Broth Base

Product Code. 11738153
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11738153 500 g Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11738153 Supplier BD Supplier No. 211767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with Fraser Broth Supplement Selective enrichment and detection of Listeria

  • Formulation: Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L

Certifications

CCAM, COMPF, ISO, USDA

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, COMPF, ISO, USDA
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fraser Broth Base
Product Type Nutrient Broth
Formulation Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L
For Use With (Application) Used with Fraser broth supplement selective enrichment and detection of Listeria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.