Learn More
BD Difco™ Dehydrated Culture Media: CzaPEK-Dox Broth
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | CzaPEK-Dox Broth |
| Product Type | Microbiology Media |
| Formulation | Saccharose 30g/L, Sodium Nitrate 3g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Potassium Chloride 0.5g/L, Ferrous Sulfate 0.01g/L |
| For Use With (Application) | Cultivation of organisms capable of utilizing inorganic Nitrogen |
| Packaging Type | Bottle |
| Quantity | 500 g |