missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Brilliant Green Agar
Click to view available options
:
Brilliant Green Agar; 500g
Description
Dehydrated form for convenience of laboratorian and bioprocessing specialist
Specifications
Specifications
| Certifications/Compliance | CE, EP, SMWW |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Brilliant Green Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone 10g/L, Yeast extract 3g/L, Lactose 10g/L, Saccharose 10g/L, Sodium Chloride 5g/L, Agar 20g/L, Brilliant green 12.5mg/L, Phenol red 0.08g/L |
| For Use With (Application) | Isolation of Salmonella other than S. Typhi from feces and other material |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction