Learn More
BD Difco™ & BBL™ Manual (Second Edition)
The Manual of Microbiological Culture Media is a valuable resource for all laboratories using dehydrated, bottled, tubed and plated media. BD Difco™ & BBL™ Manual (Second Edition) includes a CD with an electronic copy for easy access and searching.
Brand:  BD 220225
Product Code. 11708313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This second edition combines the Difco™ Manual (11th edition) and the Manual of BBL™ Products and Laboratory Procedures (6th edition). Manual has the entire line of Difco and BBL media including dehydrated culture media (DCM) and prepared plated, tubed, and bottled media.
Product descriptions include
  • Medium formulation
  • Recommended quality control organisms and expected results
  • History, ingredients, and usage information
  • Catalog numbers for all packaging configurations
  • Icons denoting “official” and “standard methods” publications
Also includes flowcharts, application tables, over 400 color photographs, and CD-ROM of manual.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For laboratory use