Learn More
BD Salmonella and Shigella Typing Antisera
Product Code. 11795965
Click to view available options
Type:
Salmonella O Antiserum Group A Factors 1, 2, 12
Salmonella O Antiserum Group B Factors 1, 4, 12, 27
Salmonella O Antiserum Group B Factors 1, 4, 5, 12
Salmonella O Antiserum Group C1 Factors 6, 7
Salmonella O Antiserum Group C2 Factors 6, 8
Salmonella O Antiserum Group C3 Factors 8
Salmonella O Antiserum Group E2 Factors 3, 15
Salmonella O Antiserum Group E3 Factors (3), (15), 34
Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37)
Salmonella O Antiserum Group I Factor 16
Salmonella O Antiserum Group K Factor 18
Salmonella O Antiserum Poly A-I Factors 1-16
Salmonella Vi Antiserum
Shigella Antiserum Poly Group C
Shigella Poly Group C1
Shigella Poly Group C2
Unit Size:
3mL
Each
Product Code. 11795965
Brand: BD 229501
Specifications
Clia Complexity High
Detectable Analytes Salmonella
Type Salmonella O Antiserum Group C2 Factors 6, 8
Quantity 3 mL
Form Lyophilized
compliance-icons
Product Identifier
  • vial sal O antis group C2
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium azide