Learn More
BD Salmonella and Shigella Typing Antisera
Click to view available options
Type:
Salmonella O Antiserum Group A Factors 1, 2, 12
Salmonella O Antiserum Group B Factors 1, 4, 12, 27
Salmonella O Antiserum Group B Factors 1, 4, 5, 12
Salmonella O Antiserum Group C1 Factors 6, 7
Salmonella O Antiserum Group C2 Factors 6, 8
Salmonella O Antiserum Group C3 Factors 8
Salmonella O Antiserum Group E2 Factors 3, 15
Salmonella O Antiserum Group E3 Factors (3), (15), 34
Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37)
Salmonella O Antiserum Group I Factor 16
Salmonella O Antiserum Group K Factor 18
Salmonella O Antiserum Poly A-I Factors 1-16
Salmonella Vi Antiserum
Shigella Antiserum Poly Group C
Shigella Poly Group C1
Shigella Poly Group C2
Unit Size:
3mL
Each
Specifications
Specifications
| Clia Complexity | High |
| Detectable Analytes | Salmonella |
| Type | Salmonella O Antiserum Group B Factors 1, 4, 5, 12 |
| Quantity | 3 mL |
| Form | Lyophilized |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
- MIXTURE LIST-Contains : Sodium Azide