Learn More
BD Cytometric Bead Array Th1/Th2 Cytokine Lyophilized Standard
Product Code. 15896698
Click to view available options
Quantity:
0.2 mL
Unit Size:
Each
Product Code. 15896698
Brand: BD 551810
Designed for use as a standard control with CBA Human Th1/Th2 Cytokine Kit and CBA Non-Human Primate Th1/Th2 Cytokine Kit
  • Contains a mixture of individually lyophilized recombinant proteins made up of human (IL)-2, IL-4, IL-5, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IFN-γ
  • Lyophilized and should be reconstituted and serially diluted before mixing with CBA capture beads and PE detection reagents
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Includes Contains a mixture of individually lyophilized recombinant proteins made up of Human (IL)-2, IL-4, IL-5, IL-6, IL-10, Tumor Necrosis Factor (TNF) and IFN-g
Quantity 0.2 mL
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry
Product Type Human Th1/Th2 Cytokine lyophilized standard
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Human Th1/Th2 Cytokine Lyophilized Standard
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT