Learn More
BD Cytometric Bead Array Mouse IL-6 Enhanced Sensitivity Flex Set
Product Code. 15801749
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15801749 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15801749 Supplier BD Supplier No. 562236
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Includes
Capture beads, detection reagent, standards.
For measuring mouse Interleukin-6 (IL-6) in serum and cell culture supernatant samples
  • Bead-based immunoassays
  • Bead B4
  • Also measures soluble analytes present in very low concentrations
TRUSTED_SUSTAINABILITY
Specifications
Antigen IL-6
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-6, flow cytometry
Product Type Mouse IL-6 enhanced sensitivity flex set (bead B4)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Mouse IL-6 Enhanced Sensitivity Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: CMIT/MIT