missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse IL-12p70 Enhanced Sensitivity Flex Set

Product Code. 15831769
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15831769 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15831769 Supplier BD Supplier No. 562264
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Bead-based immunoassay capable of measuring mouse Interleukin-12 p70 (IL-12p70) in serum and cell culture supernatant sample

Contains one vial each of Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) and two vials of Standard. The Enhanced Sensitivity Detection Reagent (Part B) is provided in the CBA Mouse Enhanced Sensitivity Master Buffer Kit. The Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) components of this flex set have been formulated to a 20× concentration to ensure product performance when multiplexed.

  • Capable of measuring soluble analytes present in very low concentrations
  • Must be used in conjunction with CBA Mouse Enhanced Sensitivity Master Buffer Kit, flow cytometer, and the FCAP Array Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12p70
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-12 p70, flow cytometry
Product Type Mouse IL-12p70 enhanced sensitivity flex set (bead D7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.