missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Flex Set Standard
Click to view available options
Description:
Human IFN-gamma Flex Set Standard
Human IL-10 Flex Set Standard
Human IL-1beta Flex Set Standard
Human IL-2 Flex Set Standard
Human IL-4 Flex Set Standard
Human IL-6 Flex Set Standard
Human IL-8 Flex Set Standard
Human TNF Flex Set Standard
Unit Size:
Each
Description
This product contains lyophilized recombinant human Interferon-λ (IFN-λ)
- Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
Flow cytometry
Specifications
Specifications
| Content And Storage | 4°C |
| Description | Human TNF Flex Set Standard |
| Format | Lyophilized |
| Includes | Lyophilized recombinant Human Interferon-l (IFN-l) |
| Quantity | 0.2 mL |
| Regulatory Status | RUO |
| Product Type | Cytometric Bead Array (CBA) Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction