missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Human IL-12 p70 Flex Set Standard
Description
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Concentration of 2500pg/mL when reconstituted in 4.0 mL Assay Diluent
- Recombinant
- Regulatory status: RUO
Flow cytometry
Order Info
One vial.
Specifications
Specifications
| Antigen | IL-12B, IL-12A |
| Cross Reactivity | Human |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-12 p70 flex set standard |
Safety and Handling
Product Identifier
- Human IL-12p70 Standard
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: CMIT/MIT
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?