missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Rat IFN-γ; Flex Set, Bead A6
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Rat IFN-γ; Standard; Rat IFN-γ; PE Detection Reagent; Rat IFN-γ; Capture Bead A6
The BD CBA Rat IFN-γ Flex Set is a bead-based immunoassay capable of measuring rat interferon-γ (IFN-γ) in serum and cell culture supernatant samples. Rat reactivity was determined by testing samples with the BD CBA Rat IFN-γ Flex Set.
- Reactivity: Rat
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow cytometry and for detection of Rat interferon |
| Product Type | Rat IFN-gamma Flex set (bead A6) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction