missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-13 Enhanced Sensitivity Flex Set (Bead B8)

Product Code. 15871769
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871769 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15871769 Supplier BD Supplier No. 562273
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Measures mouse Interleukin-13 (IL-13) in cell culture supernatant samples

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 200,000 fg/ml (when reconstituted in 0.4 ml Assay Diluent)
  • Capable of measuring soluble analytes present in very low concentrations

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-13, flow cytometry
Product Type Mouse IL-13 enhanced sensitivity flex set (bead B8)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Mouse IL-13 Enhanced Sensitivity Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: CMIT/MIT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.