missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-12p70; Flex Set, Bead D7

Product Code. 10688514
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10688514 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10688514 Supplier BD Supplier No. 558303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Mouse IL-12p70; Standard; Mouse IL-12p70; PE Detection Reagent; Mouse IL-12p70; Capture Bead D7

For detecting mouse interleukins in serum and cell culture samples

The BD CBA Mouse IL-12p70 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-12p70 (IL-12p70) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with BD CBA Mouse IL-12p70 Flex Set.

  • Reactivity: Mouse
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12p70
Cross Reactivity Mouse
Includes Mouse IL-12p70, standard, Mouse IL-12p70, PE detection reagent, Mouse IL-12p70, capture bead D7
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IL-12p70 flex set (bead D7)
compliance-icons
Product Identifier
  • Mouse IL-12p70 Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: CMIT/MIT

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.