missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human LTα; Flex Set, Bead D5
Description
Includes
Human LTα, Standard; PE Detection Reagent; Capture Bead D5.
- Bead-based immunoassay
- Formerly known as tumor necrosis factor-β
- Human reactivity determined by testing samples with BD CBA Human LT-α Flex
Specifications
Specifications
| Antigen | LT-alpha |
| Cross Reactivity | Human |
| Includes | Human LTa, standard, PE detection reagent, capture bead D5 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human LT-alpha flex set (bead D5) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?