Learn More
BD Cytometric Bead Array (CBA) Human IL-9; Flex Set, Bead B6
Product Code. 15899308
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899308 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15899308 Supplier BD Supplier No. 558333
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Includes
Human IL-9; Standard; Human IL-9; PE Detection Reagent; Human IL-9; Capture Bead B6
For detecting human interleukins in serum, plasma, and cell culture samples
The BD CBA Human IL-9 Flex Set is a bead-based immunoassay capable of measuring human interleukin-9 (IL-9) in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human IL-9 Flex Set.
  • Reactivity: Human
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Antigen IL-9
Cross Reactivity Human
Includes Human IL-9, standard, Human IL-9, PE detection reagent, Human IL-9, capture bead B6
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human IL-9 flex set (bead B6)
compliance-icons
Product Identifier
  • Human IL-9 Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: CMIT/MIT