missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IL-6; Flex Set, Bead A7

Product Code. 15829278
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829278 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829278 Supplier BD Supplier No. 558276
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Human IL-6 Standard; Human IL-6 PE Detection Reagent; Human IL-6 Capture Bead A7

Bead-based immunoassay capable of measuring human interleukin-6 (IL-6) in serum, plasma, and cell culture supernatant samples

Reactivity

  • QC Testing: Human
  • Testing in Development: Nonhuman Primate

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Cross Reactivity Human, Rhesus Monkey, Cynomolgus Monkey
Includes Human IL-6 Standard, Human IL-6 PE Detection Reagent, Human IL-6 Capture Bead A7
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human IL-6 Flex Set (Bead A7)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.