Learn More
BD Cytometric Bead Array (CBA) Human IL-3; Flex Set, Bead D5
Description
Includes
- Human IL-3 Standard
- Human IL-3, PE Detection Reagent
- Human IL-3, Capture Bead D5
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. Capture Bead and PE Detection Reagent components of this flex set have been formulated to 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 2,500pg/mL.
- Human reactivity was determined by testing samples with CBA Human IL-3 Flex Set
- Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer and CAP Array Software
Specifications
Specifications
| Antigen | IL-3 |
| Cross Reactivity | Human |
| Includes | Human IL-3 standard, Human IL-3, PE detection reagent, Human IL-3, capture bead D5 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-3 flex set (bead D5) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.