Learn More
BD Cytometric Bead Array (CBA) Human CD62P (P-Selectin); Flex Set, Bead D7
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human CD62P (P-Selectin); Standard; Human CD62P (P-Selectin); PE Detection Reagent; Human CD62P (P-Selectin); Capture Bead D7
Human reactivity was determined by testing samples with the BD CBA Human Soluble CD62P (P-Selectin) Flex Set.
- Format: Purified
Specifications
Specifications
| Antigen | CD62P (P-Selectin) |
| Cross Reactivity | Human |
| Includes | Capture bead, detection reagent and standard |
| Quantity | 100 Tests |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human CD62P (P-Selectin) flex set (bead D7) |