Learn More
BD CBA Mouse Inflammation Kit
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
Description
Includes
Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-γ, TNF, and IL-12p70; wash buffer; assay diluent; cytometer setup reagents
- Can be used to quantitatively measure Interleukin-6 (IL-6), Interleukin-10 (IL-10), Monocyte Chemoattractant Protein-1 (MCP-1), Interferon-γ (IFN-γ), Tumor Necrosis Factor (TNF), and Interleukin-12p70 (IL-12p70) protein levels in a single sample
- Detection reagent provided in the kit is a mixture of phycoerythrin (PE)-conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
- Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Mouse |
| Includes | Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-g, TNF and IL-12p70, Wash Buffer, assay diluent, cytometer setup reagents |
| Quantity | 80 Tests |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse inflammation kit |