Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Nalidixic Acid
Product Code. 11742954
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11742954 10/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742954 Supplier BD Supplier No. 231311
 Restricted Item
Estimated Shipment: 14-10-2026
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Nalidixic Acid
Concentration 30 μg
Packaging Type Disc
Quantity 10/Pk.
compliance-icons
Product Identifier
  • Disc Nalidixic Acid Na-30 10 Ea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Nalidixic acid