missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Ampicillin4, AM-10
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 500
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Specifications
| Antimicrobial Agents | Ampicillin |
| Concentration | 10 μg |
| Packaging Type | Disc |
| Quantity | 10/Pk. |
Safety and Handling
Product Identifier
- BD BBL Sensi-Disc Ampicillin Am-10 1 Ea
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ampicilin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction