missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Broth
Click to view available options
Quantity:
5 mL, 10/Pk.
5mL, Carton of 100 Size K Tubes
Media Type:
Brain Heart Infusion Broth
Unit Size:
Pack of 10
Pack of 100
Description
- For cultures requiring prolonged incubation
- Contains 6.5% sodium chloride that differentiate enterococci from nonenterococcal group D streptococci
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA |
| Color | Clear |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Brain Heart Infusion Broth |
| Formulation | Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Pancreatic digest of Gelatin 14.5g/L, Disodium Phosphate 2.5g/L |
| For Use With (Application) | Cultivation of fastidious and non-fastidious microorganisms |
| Packaging Type | K Tube |
| Quantity | 5 mL, 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction