Learn More
BD BBL™ Mycoflask™ Prepared Pour Tubed Media: Trypticase™ Soy Agar (Blood Agar Base) Deeps
Click to view available options
:
Trypticase soy agar (blood agar base) deeps; 20mL
Description
- For cultures requiring prolonged incubation
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
| Color | Beige |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Trypticase soy agar |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | Isolation and cultivation of non-fastidious and fastidious microorganisms |
| Packaging Type | Tube |
| Quantity | 20 mL |