Learn More
BD BBL™ Mycoflask™ Middlebrook 7H9 Broth with Glycerol Prepared Media
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, SMWW |
| Color | Clear |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Middlebrook 7H9 Broth |
| Formulation | Monopotassium Phosphate 2g/L, Disodium Phosphate 1.5g/L, Monosodium Glutamate 0.5g/L, Sodium Citrate 0.1g/L, Ammonium Sulfate 0.5g/L, Pyridoxine 0.001g/L, Ferric Ammonium Citrate 0.04g/L, Magnesium Sulfate 0.05g/L, Zinc Sulfate 0.001g/L, Copper Sulfate 0.001g/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L |
| For Use With (Application) | For growth of pure cultures of mycobacteria |
| Packaging Type | Tube with Tight Cap |
| Quantity | 5 mL |
For in vitro Diagnostic Use.