missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BCOR Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | BCL6 co-repressor |
Gene Alias | ANOP2,FLJ20285,FLJ38041,KIAA1575,MAA2,MCOPS2,MGC131961,MGC71031 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023515
|
Abnova™
FS0063.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16033515
|
Abnova™
FS0063.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
ANOP2,FLJ20285,FLJ38041,KIAA1575,MAA2,MCOPS2,MGC131961,MGC71031 | |
54880 | |
Liquid |
Tube | |
BCL6 co-repressor | |
BCOR | |
DNA/RNA |