Learn More
Abnova™ BCOR Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FS0063.200uL
Detalles adicionales : Weight : 0.02000kg
Especificaciones
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
ANOP2,FLJ20285,FLJ38041,KIAA1575,MAA2,MCOPS2,MGC131961,MGC71031 | |
54880 | |
200 μL |
Tube | |
BCL6 co-repressor | |
BCOR | |
DNA/RNA | |
Liquid |