missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford BCIP P-Toludine Salt
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: 5-Bromo-4-Chloro-3-Indolyl-Phospate-P-Toluidine Salt
- CAS Number: 6578-06-9
- Molecular Formula: C8H5BrCINO4P C7H10N
- Molecular Weight: 433.6
Specifications
Specifications
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction